Value Lab Upright Freezers
Thermo Scientific™

Value Lab Upright Freezers

Meet routine laboratory storage requirements with upright freezers that feature digital controls and excellent temperature stability.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)Electrical RequirementsType
02LFEETSA1.8 cu. ft.115 V 60 HzGeneral Purpose Freezer
04LFEETSA3.5 cu. ft.115 V 60 HzGeneral Purpose Freezer
Catalog number 02LFEETSA
Price (INR)
-
Capacity (English):
1.8 cu. ft.
Electrical Requirements:
115 V 60 Hz
Type:
General Purpose Freezer

Meet routine laboratory storage requirements with Thermo Scientific™ Value Lab Upright Freezers. Freezers feature digital controls and excellent temperature stability.

  • 20 cu. ft. upright
  • Four fixed wire shelves and six door shelves
  • Manual defrost
  • Enameled steel interior and exterior
  • CFC-free insulation and coolant
  • Warranty varies by country, please contact us for details
Specifications
Access SecurityDoor Lock
Amperage1.2 A
CastersNo Leveling Feet
ColorWhite
Control TypeMechanical Dial Thermostat (Outside Rear Bottom Left)
DefrostManual
Dimensions (D x W x H) Interior13 x 13.38 x 18.63 in. (33.02 x 34 x 47.32 cm)
Dimensions (L x W x H) Exterior17.75 x 18.5 x 24.5 in. (45.09 x 47 x 62.23 cm)
DisplayTemperature Display - None
Door StyleHinged
Electrical Requirements115 V 60 Hz
Exterior FinishEnameled Steel
For Use With (Application)Indoor General Storage Of Non-Critical Samples And Reagents
Frequency60 Hz
Hinge SideRight
InsulationPolyurethane Foam
InteriorABS Plastic
LightingInterior - no
Min. Clearance4 in. On Sides, 4 in. At Back, 4 in. On Top
Model02LFEETSA
Plug TypeNEMA 5-15 P
Rack Capacity10
RefrigerantR134a, 100% CFC Free
Shipping Weight (English)43 lb.
Shipping Weight (Metric)19.5 kg
Temperature (Setpoint)Adjustable
TypeGeneral Purpose Freezer
Typical Lifetime10 Years
WarrantyWarranty varies by country, please contact us for details
Wattage85 w
Weight (English)49 lb.
Weight (Metric)22 kg
Capacity (English)1.8 cu. ft.
Capacity (Metric)51 L
Certifications/CompliancecCSAus
CompressorBottom-mounted
Monitoring OptionsAlarm
No. of Doors1 (1 compartment)
Shelves1 Removable
StyleUndercounter
Temperature Range-12°C to -24°C
Voltage115 V
Unit SizeEach