Dionex™ IonPac™ NS1 and NG1 IC Columns
Actual product may vary
Dionex™ IonPac™ NS1 and NG1 IC Columns
Dionex™ IonPac™ NS1 and NG1 IC Columns
Thermo Scientific™

Dionex™ IonPac™ NS1 and NG1 IC Columns

Green features
Achieve high-efficiency separations of hydrophobic, ionizable compounds with these polymeric-based, reversed-phase columns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Particle SizeColumn Type
035321250 mm4 mm10 μmAnalytical
039568150 mm4 mm5 μmAnalytical
Catalog number 035321
Price (INR)
-
Length (Metric):
250 mm
Diameter (Metric):
4 mm
Particle Size:
10 μm
Column Type:
Analytical

Achieve high-efficiency separations of hydrophobic, ionizable compounds with Thermo Scientific™ Dionex™ IonPac™ NS1 10μm Analytical & Guard Columns. The columns of choice for ion-pair or ion-suppression chromatography, these polymer-based, reversed-phase columns provide excellent resolution and peak shape. Packing is a neutral, macroporous, high surface area, ethylvinylbenzene polymer cross linked with 55% divinylbenzene, making IonPac NS1 columns resistant to solvents, acids and bases while permitting the use of eluent from pH 0 to 14.

Polymer-Based Columns for Efficient Reversed-Phase Separations
  • Ideal for separation of large molecules that carry localized charges, such as surfactants
  • Can be used for traditional polymeric reversed-phase applications
  • Use with ion-pair chromatography for difficult separations
  • Available in 5 and 10μm particle sizes
Key Applications

Use IonPac NS1 columns for reliable separations of various compounds: High molecular weight aliphatic, carboxylic acids; Alkyl and aromatic sulfates and sulfonates; Quaternary ammonium ions; Water-soluble vitamins; Sulfur oxides; Metal cyanide complexes; Phenols

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Crosslinking55%DVB
Flow Rate1 mL/min
For Use With (Application)Reversed-Phase or Ion-Pair Chromatography
Green FeaturesFSC
HydrophobicityHigh
Max. Pressure4000 psi (275 bar)
Packing MaterialSpherical, Crosslinked, Macroporous Polymer
Particle Size10 μm
Solvents0 to 100% of HPLC solvents
Stationary PhaseEthylvinylbenzene/Divinylbenzene
TypeMixed Mode
Column TypeAnalytical
Diameter (Metric)4 mm
FormatStandard Bore
Length (Metric)250 mm
pH0 to 14
Unit SizeEach