Dionex™ 0.5M Sodium Bicarbonate Anion Eluent Concentrate
Thermo Scientific™
Dionex™ 0.5M Sodium Bicarbonate Anion Eluent Concentrate
Designed to aid methods development, this high quality Thermo Scientific™ Dionex™ 0.5M Bicarbonate Anion Eluent Concentrate enables concentrations to be customized as needed.
Have Questions?
Catalog NumberdimForUseWithQuantityConcentration
037163500 mL0.5 M
Catalog number 037163
Price (INR)
-
Quantity:
500 mL
Concentration:
0.5 M
Designed as an aid to methods development or for changing applications requirements, this ready-to-dilute Thermo Scientific™ Dionex™ 0.5M Bicarbonate Anion Eluent Concentrate enables you to make different sodium carbonate/bicarbonate eluent concentrations, quickly and easily. The concentrate, provided in 500mL quantities, is made with only high purity, reagent grade chemicals and water (18 MO-cm) and quality checked for ionic contamination. You can use this concentrate in conjunction with the Dionex 0.5M Carbonate Eluent Concentrate for custom eluent concentrations.
  • Only high-purity, reagent-grade chemicals and water (18 MΩ-cm), quality-control checked for ionic contamination, are used
  • All standards are NIST™ traceable
Includes:
A Certificate of Analysis is provided with every standard and chelation chromatography reagent
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceNIST Traceable
Concentration0.5 M
DescriptionSodium Bicarbonate Concentrate
HazardsIrritant
IncludesCertificate of Analysis
Quantity500 mL
Standard Concentration0.5 M
StorageRoom Temperature
TypeEluent Concentrate
Unit SizeEach 1