QMS™ Therapeutic Drug Monitoring (TDM) Calibrators
Actual product may vary
QMS™ Therapeutic Drug Monitoring (TDM) Calibrators
QMS™ Therapeutic Drug Monitoring (TDM) Calibrators
Thermo Scientific™
QMS™ Therapeutic Drug Monitoring (TDM) Calibrators
Accurately measure therapeutic drug levels in serum and plasma with Thermo Scientific™ QMS™ TDM Calibrators, offered in Levels A to F.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDetectable Analytes
0373787Lamotrigine
0374157Amikacin
0374652Teicoplanin
0374165Quinidine
0374173Topiramate
0373597Vancomycin
0373381Zonisamide
0373902Gentamicin
Catalog number 0373787
Price (INR)
-
Detectable Analytes:
Lamotrigine
Accurately measure therapeutic drug levels in serum and plasma with Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Calibrators, which employ advanced microparticle technology that is easy-to-use.
Reduces Labor
  • Significantly reduces labor, time, and expense compared to HPLC or LCMS and other technologies
  • Based on competitive inhibition immunoassay principle whereby free-drug in the sample competes for antibody binding sites with drug molecules coated onto uniform microparticles
Ready-to-use
  • Liquid stable and ready-to-use
  • Applicable to general chemistry analyzers
  • Validated parameters assure lab-to-lab reproducibility
  • Superb reagent stability
Optimal Sensitivity
  • Excellent correlation to reference methods
  • Low cross-reactivity with metabolites and commonly co-administered drugs
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
QuantityLevels A-F, 1 x 1 mL
TypeCalibrator
Detectable AnalytesLamotrigine
Sample TypePlasma, Serum
Unit SizeEach
Contents & Storage
2°C to 8°C