QMS™ Everolimus Immunoassays, Calibrators and Controls
Actual product may vary
QMS™ Everolimus Immunoassays, Calibrators and Controls
QMS™ Everolimus Immunoassays, Calibrators and Controls
Thermo Scientific™
QMS™ Everolimus Immunoassays, Calibrators and Controls
Obtain results you can count on for immunosuppressant drug monitoring with the Thermo Scientific™ QMS™ Everolimus Immunoassay.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeDescription
0373860Everolimus Calibrator SetFor international use only - not for sale in U.S.A.
0373852Everolimus AssayFor international use only - not for sale in U.S.A.
0373878Everolimus Control Set
Catalog number 0373860
Price (INR)
-
Type:
Everolimus Calibrator Set
Description:
For international use only - not for sale in U.S.A.
Obtain results you can always count on for the crucial monitoring of everolimus drug levels in kidney and liver transplant patients with the Thermo Scientific™ QMS™ Everolimus Immunoassay. Using our proprietary QMS technology, these liquid, ready-to-use reagents provide excellent accuracy and precision. Applications are available on our compact Indiko™ and Indiko Plus clinical and specialty chemistry systems as well as a wide variety of other laboratory instrumentation.
Key Features
  • Optimal low-end sensitivity with a broad dynamic range
  • Excellent correlation with HPLC-MS, the gold standard method
  • Precise throughout the standard assay range
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionFor international use only - not for sale in U.S.A.
Quantity6 levels, 1 x 3 mL
TypeEverolimus Calibrator Set
Detectable AnalytesEverolimus
Sample TypeWhole Blood
Unit SizeEach
Contents & Storage
2°C to 8°C