QMS™ Therapeutic Drug Monitoring (TDM) Controls
Actual product may vary
QMS™ Therapeutic Drug Monitoring (TDM) Controls
QMS™ Therapeutic Drug Monitoring (TDM) Controls
QMS™ Therapeutic Drug Monitoring (TDM) Controls
QMS™ Therapeutic Drug Monitoring (TDM) Controls
Thermo Scientific™
QMS™ Therapeutic Drug Monitoring (TDM) Controls
Ensure accurate results when testing with the newest TDM reagents with Thermo Scientific™ QMS™ TDM Controls, offered in three levels.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
0374090Lamotrigine Controls
0373373Zonisamide Controls
0374660Teicoplanin Controls
0374181Topiramate Controls
Catalog number 0374090
Price (INR)
-
Description:
Lamotrigine Controls
Ensure accurate results when testing with the newest TDM reagents with Thermo Scientific™ QMS™ TDM Controls, offered in three levels.
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration2.0, 15.0, 25.0 μg/mL
DescriptionLamotrigine Controls
LevelLevel 3
Quantity3 x 2.5 mL
TypeControl
Detectable AnalytesLamotrigine
Sample TypePlasma, Serum
Unit SizeEach
Contents & Storage
2°C to 8°C