You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
Home
▸
Shop All Products
▸
Research and Clinical Diagnostic Tests and Controls
▸
Allergy, Arthritis and Immunology Testing
▸
Immunosuppressant Drug Testing Controls and Calibrators
▸
QMS™ Therapeutic Drug Monitoring (TDM) Controls
Actual product may vary
Thermo Scientific™
QMS™ Therapeutic Drug Monitoring (TDM) Controls
Ensure accurate results when testing with the newest TDM reagents with Thermo Scientific™ QMS™ TDM Controls, offered in three levels.
Have Questions?
Change view
Catalog Number
Description
0374181
Topiramate Controls
0373373
Zonisamide Controls
0374660
Teicoplanin Controls
0374090
Lamotrigine Controls
4 Options
Catalog number
0374181
Price (INR)
-
Request A Quote
Description:
Topiramate Controls
Ensure accurate results when testing with the newest TDM reagents with Thermo Scientific™ QMS™ TDM Controls, offered in three levels.
For In Vitro Diagnostic Use.
For In Vitro Diagnostic Use.
Specifications
Concentration
High, Medium, Low
Description
Topiramate Controls
Level
Level 3
Quantity
3 x 2.5 mL
Type
Control
Detectable Analytes
Topiramate
Sample Type
Plasma, Serum
Unit Size
Each
Contents & Storage
2°C to 8°C