Dionex™ OnGuard™ Workstation
Dionex™ OnGuard™ Workstation
Dionex™ OnGuard™ Workstation
Thermo Scientific™
Dionex™ OnGuard™ Workstation
Use the Thermo Scientific™ Dionex™ OnGuard™ Workstation with Dionex™ OnGuard™ II Cartridges to pretreat up to 12 samples in parallel semi-automatically prior to ion chromatography.
Have Questions?
Catalog NumberQuantity
039599#N/A
Catalog number 039599
Price (INR)
-
Quantity:
#N/A
Pretreat up to 12 samples in parallel using the Thermo Scientific™ Dionex™ OnGuard™ Workstation.
This vacuum-driven accessory is used with Dionex OnGuard™ II Cartridges to remove interfering substances from sample matrices prior to ion chromatography. With cartridges placed in the device, a stopcock valve for each cartridge controls flow into 5mL sample reservoir. With Dionex 0.5mL PolyVials and a vacuum source, the device preterits up to 12 samples. Transfer samples directly to an autosampler. Six 10mL volumetric flasks can be used as sample collection containers.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Equipment)PolyVials, Vacuum Source, 10 mL Volumetric Flasks
IncludesIndividual stopcock valves on each sample tube to allow control of individual flow rates
OperationEnables simultaneous pretreatment of multiple samples with OnGuard sample pretreatment cartridges.
Quantity#N/A
TypeOnGuard Workstation
Unit SizeEach 1