DRI™ Cotinine Drugs of Abuse Calibrators
Product Image
Thermo Scientific™

DRI™ Cotinine Drugs of Abuse Calibrators

Utilize the globally valued accuracy, precision and lot-to-lot dependability of Thermo Scientific™ DRI™ Cotinine Drugs of Abuse Calibrators.
Have Questions?
Catalog NumberQuantity
04046 x 5 mL
Catalog number 0404
Price (INR)
-
Request A Quote
Quantity:
6 x 5 mL

Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation with Thermo Scientific™ DRI™ Cotinine Drugs of Abuse Calibrators. These liquid, ready-to-use assays have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration0, 100, 250, 500, 1000, 2000 ng/mL
DescriptionDRI Cotinine Calibrators Kit
Level6
Quantity6 x 5 mL
TypeCalibrator
Detectable AnalytesCotinine
Sample TypeUrine
Unit SizeEach
Contents & Storage
2°C to 8°C