Use these sample reservoir tubes to collect samples after removal of matrix interferences on the Thermo Scientific™ Dionex™ OnGuard™ Workstation.
Have Questions?
Catalog Number
For Use With (Equipment)
041233
OnGuard Sample Preparation Station
Catalog number 041233
Price (INR)
-
For Use With (Equipment):
OnGuard Sample Preparation Station
Choose long, 0.5mL PolyVial tubes with Thermo Scientific™ Dionex™ OnGuard™ Sample Reservoirs that are used on the Dionex OnGuard Workstation to collect samples after removal of matrix interferences. Holding up to 12 Dionex OnGuard II Cartridges, the workstation is a convenient way to remove matrix interferences from up to 12 samples in parallel before ion chromatography (IC) analysis. Simply collect treated samples in these sample reservoir tubes for direct use in the Dionex AS-DV autosampler.
Specifications
Description5 cc, Pk. of 250
For Use With (Equipment)OnGuard Sample Preparation Station