Barnstead™ Classic Stills Replacement Consumables
Actual product may vary
Thermo Scientific™
Barnstead™ Classic Stills Replacement Consumables
Maintain the performance of your Thermo Scientific™ Barnstead™ Classic Still with replacement consumables such as UV lamps and air filters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
04141Replacement UV LampBarnstead Classic Stills Reservoirs, H4005
25001Ventguard Filter ElementBarnstead Classic Stills
H3130Replacement Water SealBarnstead Classic Stills
Catalog number 04141
Price (INR)
-
Type:
Replacement UV Lamp
For Use With (Equipment):
Barnstead Classic Stills Reservoirs, H4005
Maintain the performance of your still with Thermo Scientific™ Barnstead™ Classic Stills Replacement Consumables, including UV lamp and Thermo Scientific™ Ventgard™ filter element (the consumable portion of the Ventgard filter assembly). Prevent air from entering the reservoir via the condenser atmospheric vent and protect stored water against airborne particulates, organics and CO2 with the Ventgard air filter.
Maintain the performance of your still with Thermo Scientific™ Barnstead™ Classic Stills Replacement Consumables, including UV lamp and Thermo Scientific™ Ventgard™ filter element (the consumable portion of the Ventgard filter assembly). Prevent air from entering the reservoir via the condenser atmospheric vent and protect stored water against airborne particulates, organics and CO2 with the Ventgard air filter. 
Specifications
For Use With (Equipment)Barnstead Classic Stills Reservoirs, H4005
TypeReplacement UV Lamp
Unit SizeEach