OmniPac™ PCX-500 HPLC Columns
Actual product may vary
Thermo Scientific™

OmniPac™ PCX-500 HPLC Columns

Green features
Separate cationic and neutral species in a single run using this solvent-compatible mixed-mode column.
Have Questions?
Catalog NumberLength (Metric)
04219550 mm
Catalog number 042195
Price (INR)
-
Length (Metric):
50 mm

Separate cationic and neutral analytes in a single run by using Thermo Scientific™ OmniPac™ PCX-500 Columns. With cation-exchange and reversed-phase properties, this polymer-based mixed-mode column is solvent compatible and stable over the entire pH range of 1-14. Solvents may be used to affect selectivity as well as to remove contaminants to extend column lifetime.

This column is ideal for analyzing a wide range of compounds including adrenergics, alcohols, anilines, antidepressants, anti-inflammatory agents, plating bath brighteners, cephalosporins, stains and dyes, herbicides, nucleotides and nucleosides, peptides, sulfonamides, and water-soluble vitamins.
  • Macroporous hydrophobic core provides reversed-phase retention and is coated with an cation-exchange latex bead layer for cation-exchange retention
  • Ion exchange capacity of approximately 120 μeq/column
  • 100% solvent-compatible and should always be run with at least 1% organic solvent in the mobile phase
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Application)Reversed Phase and Ion Exchange
Green FeaturesFSC
Max. Pressure4000 psi (275 bar)
Packing MaterialSpherical, Microporous Polymer
Particle Size8.5 μm
Pore Size1200 Å
QuantityEach
Stationary PhaseSulfonic Acid
USP TypeL50
Column TypeMixed Mode
Diameter (Metric)4 mm
Length (Metric)50 mm
pH0 to 14
Unit SizeEach