Dionex™ IonPac™ AS4A-SC IC Columns
Actual product may vary
Dionex™ IonPac™ AS4A-SC IC Columns
Dionex™ IonPac™ AS4A-SC IC Columns
Thermo Scientific™

Dionex™ IonPac™ AS4A-SC IC Columns

Green features
Low capacity anion-exchange column for fast, isocratic separation of inorganic anions such as nitrate or sulfate using a simple carbonate/bicarbonate eluent.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Column Type
043125250 mm2 mmAnalytical Column
04317550 mm4 mmGuard Column
04312650 mm2 mmGuard Column
Catalog number 043125
Price (INR)
-
Length (Metric):
250 mm
Diameter (Metric):
2 mm
Column Type:
Analytical Column
For fast, isocratic separation of inorganic anions in environmental waters, the Thermo Scientific™ Dionex™ IonPac™ AS4A-SC column was the preferred column for many years. It was first introduced in 1984 and is the specified column in U.S. EPA Method 300.0 (A). However, the advanced, high-capacity Dionex IonPac AS22 Carbonate Eluent Anion-Exchange column is recommended for fast analysis of inorganic anions in diverse matrices when using carbonate eluents.
Fast Isocratic Separation of Inorganic Anions in Simple Matrices
  • Compliance monitoring of common anions in drinking water as specified in U.S. EPA Methods 300.0 (A)
  • Separation of common inorganic anions in 10 minutes in water, food and beverages, and other diverse sample matrices
  • Compatible with sodium tetraborate gradient to optimize difficult separations
  • Solvent compatibility for easy column cleanup after the analysis of samples with hydrophobic components
Multiple Formats Available for Application Flexibility
  • 4×250mm, 2x250mm analytical columns
  • 4×50mm, 2x50mm guard columns
  • Longer 250mm columns facilitate higher resolution separations of more complex sample matrices
  • Smaller i.d. columns provide higher mass sensitivity, eluent and reagent conservation and lower operating costs

Note: Use with the Dionex IonPac AS4A Eluent Concentrate for simple, accurate eluent preparation.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceMeets or Exceeds US EPA Method 300.0 (A) Performance Requirements
Column Capacity5 μeq
Column ChemistryCarbonate/Bicarbonate
Column ConstructionPEEK
Crosslinking0.5% DVB
Flow Rate0.5 to 0.75 mL/min.
For Use With (Application)Separation of Inorganic Anions, Anion Exchange
For Use With (Equipment)ASRS Anion Self-Regenerating Suppressor
Green FeaturesFSC
HydrophobicityMedium-Low
Ion Exchange GroupAlkanol Quaternary Ammonium Ion
Max. Pressure4000 psi (275 bar)
Packing MaterialEthylvinylbenzene/Divinylbenzene
Particle Size13 μm
QuantityEach 1
Solvent CompatibilitypH 0 to 14, 0 to 100% HPLC Solvents
Solvents0 to 100% of HPLC Solvents
Stationary PhaseIonPac AS4A-SC
Column TypeAnalytical Column
Diameter (Metric)2 mm
Length (Metric)250 mm
pH0 to 14
Unit SizeEach 1