Dionex™ CTC Cation Trap Columns
Actual product may vary
Thermo Scientific™

Dionex™ CTC Cation Trap Columns

Green features
Remove trace cation contaminants from eluents using these high-capacity, low-efficiency, cation-exchange trap columns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Description
04313235 mm4 mmCTC
04019224 mm9 mmCTC-1
07901935 mm4 mmCTC 500
07597724 mm9 mmCTC 500
Catalog number 043132
Price (INR)
-
Length (Metric):
35 mm
Diameter (Metric):
4 mm
Description:
CTC

Remove trace cation contaminants from eluents using the Thermo Scientific™ Dionex™ IonPac™ CTC-1 and Dionex IonPac CTC 500 Cation Trap Columns. These high-capacity, low-efficiency, cation-exchange trap columns are installed in the eluent line after the pump prior to the injection valve to prevent spurious peaks and reduce baseline shifts during gradient chromatography. The Dionex IonPac CTC 500 is identical to the Dionex IonPac CTC-1, but is packed in a column body that is compatible with nonstop operation of high-pressure Thermo Scientific™ Dionex™ HPIC™ system pressures up to 5000 psi (34.5 MPa). For Reagent-Free™ ion chromatography (RFIC™) MSA eluent generation applications, use the Dionex CR-CTC.

Engineered for Superior Chromatographic Performance
  • High-capacity, low-efficiency cation-exchange resin in the hydronium form
  • 3.0meq/column capacity (4mm)
  • 0.8meq/column capacity (2mm)
  • 4 ' 35mm format for 2mm and 3mm applications
  • 9 ' 24mm format for 4mm applications
  • Periodic off-line regeneration extends usage
About Anion Trap Columns

The Dionex IonPac CTC-1 and Dionex IonPac CTC 500 Cation Trap Columns contain microporous cation-exchange resin in the hydronium form. The cation trap columns strip trace cationic contaminants out of the eluent and/or deionized water and prevents them from reaching the guard or analytical columns. Cationic contaminants present on the eluent and/or deionized water often interfere with the precision of trace cation determinations in particular when performing an cation-exchange application that involves a gradient eluent.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column Capacity0.8 meq
Crosslinking8% DVB
DescriptionCTC
For Use With (Application)Removal of Trace Cation Contamination from Eluents
For Use With (Equipment)2 mm and 3 mm Cation Exchange Columns
Green FeaturesFSC
HydrophobicityLow
Max. Pressure3000 psi (207 bar)
Particle Size500 μm
QuantityEach 1
Solvent CompatibilityAll Cation Exchange Eluents, 0 to 50% HPLC Solvents
Solvents0 to 50% HPLC Solvents
Stationary PhaseIonPac CTC-1 Cation
Column TypeCation Trap Column
Diameter (Metric)4 mm
Length (Metric)35 mm
Unit SizeEach 1