Dionex™ ED50A Electrochemical Detector Working Gold Electrode, with gasket and polishing pad
Offer consistent results with very good lifetimes with Thermo Scientific™ Dionex™ ED50A Electrochemical Detector Working Electrodes.
Have Questions?
Catalog number 044112
Price (INR)
-
Offer consistent results with very good lifetimes with Thermo Scientific™ Dionex™ ED50A Electrochemical Detector Working Electrodes.
Electrodes consist of a rod of the electrode material embedded in a polymeric support
CWEs provide users with the option to polish electrode surfaces should the performance degrade over time or when performance diminishes as a result of multiple analyses
Four types of electrode materials are available
Gold
Silver
Platinum
Glassy carbon
Specifications
DescriptionED50 A Working Electrode, Au
Includeswith gasket and polishing pad
MaterialGold in a Polymeric Support
For Use With (Equipment)ED50 Electrochemical Detector