Dionex™ ED50A Electrochemical Detector pH Reference Electrode
Thermo Scientific™
Dionex™ ED50A Electrochemical Detector pH Reference Electrode
The ED50A Electrochemical Detector pH Reference Electrode features a combination Detector pH-Ag/AgCl Reference Electrode which can be used for carbohydrate and amino acid analysis.
Have Questions?
Catalog NumberFor Use With (Equipment)
044198Electrochemical Detector
Catalog number 044198
Price (INR)
-
For Use With (Equipment):
Electrochemical Detector
Feature a combination Detector pH-Ag/AgCl Reference Electrode which can be used for carbohydrate and amino acid analysis with the Thermo Scientific™ Dionex™ ED50A Electrochemical Detector pH Reference Electrode.
Combination pH-Ag/AgCl electrode
  • For carbohydrate analysis, electrode is used in the Ag mode; for amino acid analyses it is used in the pH mode
Specifications
DescriptionpH Reference Electrode
For Use With (Application)Ion Chromatography
For Use With (Equipment)Electrochemical Detector
Product LineDionex
Unit SizeEach