Dionex™ ED Electrochemical Detection Disposable Electrode Gaskets
Actual product may vary
Thermo Scientific™
Dionex™ ED Electrochemical Detection Disposable Electrode Gaskets
These high-quality Thermo Scientific™ Dionex™ ED Electrochemical Detection Disposable Electrode Gaskets are for use with disposable electrodes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionMaterial
045972for ED50 A/ED50/ED40 Amperometry Cell, 1.0 mil
060141for Disposable Electrode 0.002 in.PTFE
069339for Disposable Electrode 0.001 in.ULTEM
072117for Disposable Electrode 0.001 in.PTFE
Catalog number 045972
Price (INR)
-
Description:
for ED50 A/ED50/ED40 Amperometry Cell, 1.0 mil
These high-quality Thermo Scientific™ Dionex™ ED Electrochemical Detection Disposable Electrode Gaskets are for use with disposable electrodes.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Descriptionfor ED50 A/ED50/ED40 Amperometry Cell, 1.0 mil
For Use With (Application)Ion Chromatography
QuantityEach
For Use With (Equipment)ED50A/ED50/ED40 Amperometry Cells
Product LineDionex
Unit SizeEach