Product Image
Thermo Scientific™

DRI™ Cotinine Drugs of Abuse Controls

Reduce time-consuming reagent preparation with Thermo Scientific™ DRI™ Cotinine Drugs of Abuse Controls—ready-to-use controls in lilquid form.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConcentration
0460300 ng/mL
0470700 ng/mL
Catalog number 0460
Price (INR)
-
Concentration:
300 ng/mL
Reduce time-consuming reagent preparation with Thermo Scientific™ DRI™ Cotinine Drugs of Abuse Controls—ready-to-use controls in lilquid form. Delivering high analytical accuracy, these assays have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Concentration300 ng/mL
DescriptionDRI Cotinine Drugs of Abuse Controls
Quantity1 x 5 mL
TypeControl
Detectable AnalytesCreatinine
Sample TypeUrine
Unit SizeEach