Dionex™ ASE™ 150/200/350 Collection Vials
Thermo Scientific™

Dionex™ ASE™ 150/200/350 Collection Vials

Collect extract from Thermo Scientific™ Dionex™ ASE™ 150/200/350 Accelerated Solvent Extractors using these 40mL and 60mL collection vials.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorFor Use With (Equipment)Type
048783ClearDionex ASE 200Collection Vial
048780AmberDionex ASE 200Collection Vial
048784ClearDionex ASE 150, 350, and 200Collection Vial
048781AmberDionex ASE 150, 350, and 200Collection Vial
055442
also known as 55442
ClearDionex ASE 200Graduated Concentration Vial
055444BlackDionex ASE 200, Graduated VialsSpacer
055395TranslucentASE 100, 150, 200, 300, and 350Septa
Catalog number 048783
Price (INR)
-
Color:
Clear
For Use With (Equipment):
Dionex ASE 200
Type:
Collection Vial
Collect solvent and analytes extracted from Thermo Scientific™ Dionex™ ASE™ 150/200/350 Accelerated Solvent Extractors using these collection vials. Placed in the vial tray at the lower right of the system, these vials are available in 40 and 6 0mL clear and amber glass, and are complete with caps and solvent-resistant (TFE) septa. Additional septa can also be ordered.

Compatibility

  • 40mL and 40mL graduated vials are only compatible with Dionex ASE 200 systems.
  • 40 mL collection vessels are not compatible with Dionex ASE 150 or 350 systems.
  • 60 mL collection vials require an adaptor for use with Dionex ASE 150 systems, and are not compatible with Dionex ASE 100 systems.

See Dionex™ ASE™ Collection Bottles and Lids to order 250 mL collection bottles for larger volume extraction collection.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
ColorClear
DescriptionCollection Vial
QuantityCase of 72
Volume (Metric)40 mL
For Use With (Equipment)Dionex ASE 200
TypeCollection Vial
Unit SizeCase of 72