Product Image
Thermo Scientific™
DRI™ Methaqualone Drugs of Abuse Assays
Save time in reagent preparation with Thermo Scientific™ DRI™ Methaqualone Drugs of Abuse Assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
05141 x 100 mL
05151 x 500 mL
Catalog number 0514
Price (INR)
-
Quantity:
1 x 100 mL
Save time in reagent preparation with Thermo Scientific™ DRI™ Methaqualone Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. These liquid, ready-to-use assays have been field-tested on a wide range of analyzers toeliver high analytical accuracy. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsMGC Multi-Drug Controls
DescriptionDRI Methaqualone Drugs of Abuse Assays
Quantity1 x 100 mL
Sample TypeUrine
TypeReagent
Detectable AnalytesMethaqualone
DoA CalibratorsMulti-drug Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C