Dionex™ IonPac™ CS15 IC Columns
Actual product may vary
Dionex™ IonPac™ CS15 IC Columns
Dionex™ IonPac™ CS15 IC Columns
Thermo Scientific™

Dionex™ IonPac™ CS15 IC Columns

Green features
Moderate-capacity cation-exchange column with unique selectivity for the analysis of samples with high ammonium to low ethanolamine ratios.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Column Type
052252250 mm2 mmAnalytical Column
051795250 mm4 mmAnalytical Column
05220050 mm4 mmGuard Column
05225650 mm2 mmGuard Column
Catalog number 052252
Price (INR)
-
Length (Metric):
250 mm
Diameter (Metric):
2 mm
Column Type:
Analytical Column

Obtain consistent, reliable results for the determination of disparate concentration ratios of ethanolamine and ammonium in diverse sample matrices using the Thermo Scientific™ Dionex™ IonPac™ CS15 Cation-Exchange Column. This is a medium-capacity carboxylate-functionalized cation-exchange column with crown ether moiety. The column has unique selectivity, and is the only carboxylic acid based cation column from Dionex in which ethanolamines elute before ammonium. Thus, it is the column of choice for samples with high ammonium to low ethanolamine ratios.

The Dionex IonPac CS15 column can also separate disparate ratios of sodium and ammonium, however, the advanced Thermo Scientific™ Dionex™ IonPac™ CS16 Cation Exchange column is recommended for this application.

Unique Selectivity for Ethanolamine and Ammonium in Complex Matrices
  • Medium-capacity cation exchange column designed for the determination of trace levels of ethanolamine in high levels of ammonium in complex samples
  • Also ideal for the determination of trace-level sodium in amine-treated cooling waters with resolution of ammonium and sodium ratios up to 8,000:1
  • Determination of low level cations in matrices with high concentrations of potassium
  • Matrices include environmental waters; power plant waters treated with ammonium, morpholine or ethanolamine; chemical additives; chemical process solutions; scrubber solutions; plating baths; industrial solvents; and soil matrices
  • Maximum peak efficiencies at an optimized operating temperature of 40 ˚C
  • Compatible with organic solvents, excluding alcohols, to enhance analyte solubility, modify column selectivity, and facilitate effective column clean-up
Multiple Formats Available for Application Flexibility
  • 4x250mm, 2x250mm analytical columns
  • 4x50mm, 2x50mm guard columns
  • Longer 250mm columns facilitate higher resolution separations of more complex sample matrices
  • Smaller i.d. columns provide higher mass sensitivity, eluent and reagent conservation and lower operating costs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column Capacity700 μeq
Column ChemistryAcidic
Crosslinking55% DVB
Flow Rate0.3 to 0.75 mL/min.
For Use With (Application)Cation Exchange
For Use With (Equipment)CSRS
Green FeaturesFSC
HydrophobicityMedium
Max. Pressure4000 psi (275 bar)
Packing MaterialEthylvinylbenzene/Divinylbenzene
Particle Size8.5 μm
QuantityEach 1
Solvent CompatibilityAcidic Eluents, 0 to 100% Acetonitrile, Alcohols Should be Avoided
Solvents0 to 100% Acetonitrile. Alcohols Should be Avoided.
Stationary PhaseIonPac CS15
Column TypeAnalytical Column
Diameter (Metric)2 mm
FormatMicrobore
Length (Metric)250 mm
Unit SizeEach 1