Dionex™ IonPac™ AS14A IC Columns
Actual product may vary
Dionex™ IonPac™ AS14A IC Columns
Dionex™ IonPac™ AS14A IC Columns
Thermo Scientific™

Dionex™ IonPac™ AS14A IC Columns

Green features
High capacity anion-exchange column for fast isocratic separation of inorganic anions in environmental matrices using a simple carbonate/bicarbonate eluent.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Column Type
05689930 mm3 mmGuard Column
056901150 mm3 mmAnalytical Column
056904250 mm4 mmAnalytical Column
05689750 mm4 mmGuard Column
Catalog number 056899
Price (INR)
-
Length (Metric):
30 mm
Diameter (Metric):
3 mm
Column Type:
Guard Column
Deliver fast isocratic separation of inorganic anions from diverse and complex matrices such as drinking water, wastewater, foods and beverages, and scrubber solutions with the Thermo Scientific™ Dionex™ IonPac™ AS14A Carbon Eluent Anion-Exchange Column. The Dionex IonPac AS14A column meets or exceeds the requirements of U.S EPA Method 300.0 (A). However, the Dionex IonPac AS22 Carbonate Eluent Anion-Exchange column, an advanced carbonate eluent column, is recommended for fast analysis of inorganic anions. For the fastest separation of inorganic anions using a carbonate/bicarbonate eluent, the Dionex IonPac AS22-Fast column is recommended.
Fast Separation of Inorganic Anions in Diverse Matrices
  • Compliance monitoring of common anions in drinking water as specified in U.S. EPA Method 300.0 (A)
  • Separation of common inorganic anions in 8 min (3 x 150mm format) or 13 min (4 x 250mm format) in diverse sample matrices
  • Superior retention and quantification of fluoride
  • Compatible with sodium or potassium hydroxide gradient to optimize difficult separations
  • Solvent compatibility permits easy column cleanup after the analysis of samples with hydrophobic components.
Multiple Formats Available for Application Flexibility
  • 4x250mm, 3x150mm analytical columns
  • 4x50mm, 3x30mm guard columns
  • Longer 250mm columns facilitate higher resolution separations of more complex sample matrices
  • Shorter 150mm columns produce faster runs of relatively clean matrices
  • Smaller i.d. columns provide higher mass sensitivity, eluent and reagent conservation and lower operating costs

Note: Use with the Dionex IonPac AS14 Eluent Concentrate and the Combined Seven Anion Standard for simple, accurate eluent preparation.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceU.S. EPA Method 300.0 (A)
Column Capacity8 μeq
For Use With (Application)Anion Exchange
For Use With (Equipment)IonPac AS14A Analytical Column
Green FeaturesFSC
HydrophobicityModerate
Max. Pressure4000 psi (275 bar)
Particle Size5 μm
QuantityEach 1
Solvent CompatibilitypH 0 to 14, 0 to 100% HPLC Solvents
Stationary PhaseIonPac AG14A
Column TypeGuard Column
Diameter (Metric)3 mm
FormatMicrobore
Length (Metric)30 mm
Unit SizeEach 1