Product Image
Thermo Scientific™
Dionex™ Combined Seven Anion Standard I
Increase your productivity and reduce standard preparation errors with single and combined ion standards for routine anion and cation ion chromatography determinations.
Have Questions?
Catalog NumberQuantity
056933Each 1
Catalog number 056933
Price (INR)
-
Quantity:
Each 1
Ion Standards are used for a variety of routine anion, cation, and standard determinations. Increase your productivity and reduce standard preparation errors with Thermo Scientific™ Dionex™ Ion Standards for ion chromatography. Available for routine anion and cation determinations, these single and combined ion standards are made with high purity, reagent-grade chemicals and water. Each standard is NIST traceable and includes a Certificate of Analysis.
Specifications
DescriptionDionex Combined Seven Anion Standard I, 50 mL
For Use With (Application)Routine Anion Determinations
IncludesFluoride 20 mg/L; Chloride 30 mg/L; Nitrite 100 mg/L; Bromide 100 mg/L; Nitrate 100 mg/L; Phosphate 150 mg/L; Sulfate 150 mg/L
OperationUsed for routine anion determinations.
QuantityEach 1
Standard ComponentsSodium Fluoride, Sodium Chloride, Sodium Nitrite, Sodium Bromide, Sodium Nitrate, Potassium Phosphate
Standard Concentration20, 30, 100, 100, 100, 150, 150 mg/L
StorageRoom Temperature
Volume (Metric)50 mL
TypeStandard
Unit SizeEach