Dionex™ AS14A Eluent Concentrate; Sodium Carbonate/Bicarbonate Concentrate
Actual product may vary
Thermo Scientific™

Dionex™ AS14A Eluent Concentrate; Sodium Carbonate/Bicarbonate Concentrate

For fast, reliable anion eluent preparation, this ready-to-dilute eluent concentration saves time and eliminates potential contamination or preparation error.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConcentrationQuantity
056937100 X250 mL
05756020 X100 mL
Catalog number 056937
Price (INR)
-
Concentration:
100 X
Quantity:
250 mL
For fast, reliable eluent preparation, the ready-to-dilute Thermo Scientific™ Dionex™ AS14A Eluent Concentrate; Sodium Carbonate/Bicarbonate Concentrate will save you time and eliminate potential contamination and preparation error. For use with Dionex™ IonPac™ AS14A columns, simply prepare eluent by dissolving the concentrate in deionized water. The 20x and 100x concentrate in 100mL and 250mL quantities is made with only high purity, reagent grade chemicals and water (18 MO-cm) and quality checked for ionic contamination.
Product Detail

Available in two concentrations and sizes, this concentrate is prepared easily by dissolving in deionized water;

20x Concentration (100mL)

0.80M sodium carbonate

0.10M bicarbonate concentrate

100x Concentration (250mL)

0.80M sodium carbonate

0.10M bicarbonate concentrate

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Concentration100 X
Description250 mL, (100 x)
For Use With (Equipment)IonPac AS14A Columns
HazardsIrritant
Quantity250 mL
Standard ComponentsSodium Carbonate, Sodium Bicarbonate in Deionized Water
Standard Concentration0.80, 0.10 M
StorageRoom Temperature
TypeEluent Concentrate
Unit SizeEach 1