Dionex™ ASE™ Extraction Cell Funnels
Actual product may vary
Dionex™ ASE™ Extraction Cell Funnels
Dionex™ ASE™ Extraction Cell Funnels
Dionex™ ASE™ Extraction Cell Funnels
Dionex™ ASE™ Extraction Cell Funnels
Thermo Scientific™
Dionex™ ASE™ Extraction Cell Funnels
Make it easy to add sample to the extraction cell of any Thermo Scientific™ Dionex™ ASE™ Accelerated Solvent Extractor with this Dionex ASE Extraction Cell Funnel.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
056958
also known as 56958
Dionex ASE 200 Cells
05669934, 66, and 100 mL Cells for Dionex ASE 100, 150, 300, and 350
0680755, 10, and 22 mL Cells for Dionex ASE 150 and 350
0680761 mL Cells for Dionex ASE 150 and 350
Catalog number 056958
also known as 56958
Price (INR)
-
For Use With (Equipment):
Dionex ASE 200 Cells
Make it easy to add sample to the extraction cell of any Thermo Scientific™ Dionex™ ASE™ Accelerated Solvent Extractor with this Dionex ASE Extraction Cell Funnel.
Dionex ASE systems reduce sample preparation from hours to minutes by performing automated solvent extraction on solid and semisolid sample matrices. Extraction cells feature finger-tight cell caps with compression seal for high-pressure closure and maximum ease of use.
Dionex ASE systems reduce sample preparation from hours to minutes by performing automated solvent extraction on solid and semisolid sample matrices. Extraction cells feature finger-tight cell caps with compression seal for high-pressure closure and maximum ease of use.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
MaterialAluminum
For Use With (Equipment)Dionex ASE 200 Cells
TypeFunnel
Unit SizeEach