Dionex™ DCR Displacement Chemical Regeneration Reagents
Actual product may vary
Thermo Scientific™

Dionex™ DCR Displacement Chemical Regeneration Reagents

Use these sulfuric acid and tetrabutylammonium hydroxide concentrates to prepare regenerants when operating IC system in DCR mode.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeVolume (Metric)
057559Anion Regenerant Concentrate75 mL
057555Anion Regenerant Concentrate4 x 75 mL
057556Cation Regenerant Concentrate4 x 100 mL
057561Cation Regenerant Concentrate100 mL
Catalog number 057559
Price (INR)
-
Type:
Anion Regenerant Concentrate
Volume (Metric):
75 mL
Use Thermo Scientific™ Dionex™ DCR Displacement Chemical Regeneration Reagents to prepare regenerants from concentrates when operating your ion chromatography (IC) system in DCR mode. DCR is an easy-to-use mode of operation for chemical suppressors that uses effluent from the conductivity cell to drive the regenerant into the regenerant chamber of the suppressor in a closed bottle configuration. DCR reagents are available as an anion regenerant concentrate (2N H2SO4) and a cation regenerant solution (2.06M TBAOH) in single or 4-pack bottles.
Only high-purity, reagent-grade chemicals and water (18 MO-cm), quality-control checked for ionic contamination, are used. All standards are NIST traceable.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Concentration2.0 N H2SO4
For Use With (Application)Ion Chromatography
HazardsIrritant
IncludesCertificate of Analysis
QuantityEach 1
Standard ComponentsSulfuric Acid Aqueous Solution
Standard Concentration2 N
StorageRoom Temperature
Volume (Metric)75 mL
TypeAnion Regenerant Concentrate
Unit SizeEach 1