Dionex™ Reagent-Free™ Controller DX-120 Adapter Cables
Thermo Scientific™
Dionex™ Reagent-Free™ Controller DX-120 Adapter Cables
Upgrade your legacy Thermo Scientific™ Dionex™ ion chromatography (IC) system with this required cable.
Have Questions?
Catalog NumberFor Use With (Equipment)
057861RFC-30 and RFC-10
Catalog number 057861
Price (INR)
-
For Use With (Equipment):
RFC-30 and RFC-10
Upgrade your legacy Thermo Scientific™ Dionex™ ion chromatography (IC) system with electrolytic suppression, using a Dionex RFC-10 or RFC-30 Reagent-Free™ Controller and its required cable, the Dionex DX-120 Adapter Cable.
The Dionex Reagent-Free Controller DX-120 Adapter Cable is required by the RFC-10 Reagent-Free Controller as well as for the RFC-30 Reagent-Free Controller when used with the Dionex DX-120 IC system.
Specifications
DescriptionDX-120 Adapter cable for RFC-30 series
For Use With (Equipment)RFC-30 and RFC-10
TypeAdapter Cable for RFC-30 Series
Unit SizeEach