Dionex™ Trap Column/Suppressor Cleanup Kit
Thermo Scientific™
Dionex™ Trap Column/Suppressor Cleanup Kit
Regenerate and cleanup Thermo Scientific™ Dionex™ IC trap columns and suppressors using this reagent delivery system kit.
Have Questions?
Catalog NumberFor Use With (Equipment)
059659ATC-3 and ATC-HC Anion Trap Columns
Catalog number 059659
Price (INR)
-
For Use With (Equipment):
ATC-3 and ATC-HC Anion Trap Columns
Use the Thermo Scientific™ Dionex™ Trap Column/Suppressor Cleanup Kit to deliver reagent solutions to regenerate and cleanup our trap columns and suppressors. This stand-alone pneumatically driven reagent delivery system eliminates the need for using the analytical pump for delivery of concentrated acids, bases or other reagents that can contaminate the IC system. The kit consists of a 1L plastic reservoir, a gas pressure regulator, a two-way liquid shut-off valve, and tubing and connection fittings. The kit is pre-assembled and easy to install and operate.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionTrap Column and Suppressor Regeneration Kit
For Use With (Equipment)ATC-3 and ATC-HC Anion Trap Columns
Includes1 L plastic bottle, pressure regulator, 2-way valve and fittings, and tubing
OperationFor the regeneration of the ATC-3 column.
QuantityEach 1
TypeRegeneration Kit
Unit SizeEach 1