DRI™ Methadone Drugs of Abuse Assays
Product Image
Thermo Scientific™
DRI™ Methadone Drugs of Abuse Assays
Use Thermo Scientific™ DRI™ Methadone Drugs of Abuse Assays to achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
05971 x 500 mL
05961 x 100 mL
100164033 x 18 mL
Catalog number 0597
Price (INR)
-
Request A Quote
Quantity:
1 x 500 mL
Use Thermo Scientific™ DRI™ Methadone Drugs of Abuse Assays to achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation. These liquid, ready-to-use assays offer applications for an array of clinical chemistry analyzers. Field-tested on a wide range of analyzers, these assays are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
Control SetsMGC Multi-Drug Controls
DescriptionDRI Methadone Drugs of Abuse Assays
Quantity1 x 500 mL
Sample TypeUrine
TypeReagent
Detectable AnalytesMethadone (MTD)
DoA CalibratorsMulti-drug Calibrator
Unit SizeEach
Contents & Storage
2°C to 8°C