Dionex™ AS-DV Autosampler Simultaneous Injection Kit
Thermo Scientific™
Dionex™ AS-DV Autosampler Simultaneous Injection Kit
The Thermo Scientific™ Dionex™ Simultaneous Injection Kit for ICS Systems directs samples to two independent ion chromatography (IC) systems with one AS-DV.
Have Questions?
Catalog NumberFor Use With (Equipment)
060258Ion Chromatography Systems
Catalog number 060258
Price (INR)
-
For Use With (Equipment):
Ion Chromatography Systems
Direct samples to two independent ion chromatography (IC) systems with one AS-DV, the Thermo Scientific Dionex™ AS-DV Simultaneous Injection Kit for ICS Systems.
Requires:
Chromeleon™ 6.8 SR7 (or higher)
Specifications
For Use With (Equipment)Ion Chromatography Systems
OperationDirects samples to two independent ion chromatography (IC) systems.
Required OptionsChromeleon 6.8 SR7 (or higher)
TypeInjection Kit
Unit SizeEach