Acclaim™ 300 C18 HPLC Columns
Thermo Scientific™

Acclaim™ 300 C18 HPLC Columns

Green features
Use these wide-pore C18 columns to achieve high-resolution protein separations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
06026350 mm2.1 mm
060264150 mm2.1 mm
06026550 mm4.6 mm
060266150 mm4.6 mm
063684150 mm3 mm
06969010 mm2.1 mm
06969710 mm4.6 mm
Catalog number 060263
Price (INR)
-
Length (Metric):
50 mm
Diameter (Metric):
2.1 mm
Achieve high-resolution peptide mapping and protein separations with Thermo Scientific™ Acclaim™ 300 C18 LC Columns. The 3μm ultrapure silica particles reduce the limits for diffusion of the analytes into the stationary phase, resulting in fast, high-resolution separations. Compared to 5μm column packings, the smaller particles support increased flow rates and shallower gradients on shorter columns for faster separations.
High-Resolution Separation of Proteins and Peptides
  • High-performance C18 bonding chemistry on 300Å pore silica
  • Application tested for suitability in peptide mapping
  • Reproducible for dependable results
  • Extremely low bleed, fully compatible with MS
  • Stable between pH 2 .5 and 7.5
About the Acclaim Family of HPLC Columns
  • Complementary selectivity, high column efficiencies and ultrahigh resolution
  • Broad spectrum of stationary phases, including conventional C8 and C18 phases, polar embedded phases and specialty phases including wide-pore
  • Performance tested to ensure high quality and reliability
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Column TypeReversed Phase
EndcappedYes
Flow Rate1 mL/min
For Use WithLC/MS
Green FeaturesFSC
Max. Pressure4500 psi (310 bar)
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
QuantityEach
Stationary PhaseC18
Surface Area100 m²/g
Temperature60°C
USP TypeL1
Carbon Load8%
Diameter (Metric)2.1 mm
Length (Metric)50 mm
MaterialGlass Lined Tubing
Particle Size3 μm
PhaseReversed Phase
Pore Size300 Å
Product LineAcclaim
TechniqueBio-LC
TypeAnalytical Column
pH2.5 to 7.5
Unit SizeEach