Product Image
Thermo Scientific™
Dionex™ ED50A Electrochemical Detector Cells
Detect a variety of electro-active species using ED50A Electrochemical Detector Cells.
Have Questions?
Catalog NumberFor Use With (Equipment)
060358LC10/20/30
Catalog number 060358
Price (INR)
-
For Use With (Equipment):
LC10/20/30
Detect a variety of electro-active species using Thermo Scientific™ Dionex™ ED50A Electrochemical Detector Cells.
Detects carbohydrates, amino acids (on a gold working electrode), cyanide, bromide, iodide, thiocyanate, sulfide, thiosulfate (on a silver electrode), alcohols, aldehydes, ketones, carboxylic acids (on a platinum electrode) and catecholamines. Also, detects S-containing amino acids and electroactive DNA bases and derivatives (on a carbon electrode).
Specifications
For Use With (Equipment)LC10/20/30
OperationUsed for the detection of a variety of electro-active species.
TypeElectrochemical Detector Cell
Unit SizeEach