DNAPac™ PA200 Oligonucleotide HPLC Columns
Thermo Scientific™

DNAPac™ PA200 Oligonucleotide HPLC Columns

Green features
Enjoy strong anion exchange for high-resolution analysis and purification of synthetic oligonucleotides with Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Particle Size
063421250 mm9 mm8 μm
063000250 mm4 mm8 μm
063425250 mm2 mm8 μm
06299850 mm4 mm8 μm
06341950 mm9 mm8 μm
06342350 mm2 mm8 μm
082510250 mm4.6 mm4 μm
082509150 mm4.6 mm4 μm
08250850 mm4.6 mm4 μm
088781250 mm22 mm8 μm
08878050 mm22 mm8 μm
Catalog number 063421
Price (INR)
-
Length (Metric):
250 mm
Diameter (Metric):
9 mm
Particle Size:
8 μm

Enjoy strong anion exchange for high-resolution analysis and purification of synthetic oligonucleotides with Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns.

  • Achieve n, n-1 resolution for oligonucleotides
  • Resolve oligonucleotides with secondary structures
  • Assay phosphorothioate purity
  • Selectivity control with eluent pH, salt, and solvent
  • Resolve RNA with aberrant (2' – 5') links from normal SSRNA
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Column TypeIon Exchange
Flow Rate1.2 mL/min
For Use WithUltiMate 3000 Titanium System
Green FeaturesFSC
Max. Pressure4000 psi (275 bar)
Mobile Phase CompatibilitypH 2 to 12.5, 100% compatible with common organic solvents, Ionic form eluents: chloride, perchlorate
Packing MaterialSpherical, Nonporous Polymer
QuantityEach
Stationary PhaseQuaternary Ammonium
Temperature85°C
Diameter (Metric)9 mm
Length (Metric)250 mm
Particle Size8 μm
PhaseIon Exchange
Pore SizeNonporous
Product LineDNAPac
TechniqueBio-LC
TypeAnalytical Column
pH2.5 to 12.5
Unit SizeEach