Dionex™ AS-HV Autosampler Injection 6-Port Valve
Thermo Scientific™
Dionex™ AS-HV Autosampler Injection 6-Port Valve
The MX Series II automated fluidic valve provides added flexibility where stand-alone injection is required.
Have Questions?
Catalog NumberFor Use With (Equipment)
063573AS-HV Autosampler
Catalog number 063573
Price (INR)
-
For Use With (Equipment):
AS-HV Autosampler
The Thermo Scientific™ MX Series II automated fluidic valve provides added flexibility where stand-alone injection is required. Easily combine the MX with the valves in your existing instrument to support complex fluid switching and sample injection needs.
Specifications
For Use With (Equipment)AS-HV Autosampler
Injection TypeAutomated
Product Type6-Port Injection Valve
Unit SizeEach