Dionex™ AXP Auxiliary Pump
Thermo Scientific™

Dionex™ AXP Auxiliary Pump

The Thermo Scientific™ Dionex™ AXP Auxiliary Pump is a single-piston, isocratic metering pump for ion chromatography sample and reagent delivery.
Have Questions?
Catalog NumberFor Use With (Equipment)
063973ICS-5000
Catalog number 063973
Price (INR)
-
For Use With (Equipment):
ICS-5000

The Thermo Scientific™ Dionex™ AXP Auxiliary Pump is a single-piston, isocratic metering pump for ion chromatography sample and reagent delivery.

  • Chemically inert, metal-free, PEEK™-based flow path eliminates corrosion and contamination problems
  • Digital control and status
  • Wide flow rate range
  • Internal pulse damper
  • Compatible with IC and bio applications
  • Standard positive seal washing
  • Flexible, advanced Chromeleon™ control
Specifications
DescriptionAuxiliary pump
Electrical Requirements120 to 230 VAC 50/60 Hz
For Use With (Equipment)ICS-5000
Frequency50/60 Hz
MaterialPEEK, ceramic and inert polymers
Min. Flow Rate0.01 mL/min.
PC ConnectionUSB
Pressure Ripple2% peak-to-peak (at 100 psi, 1 mL/min with in-line pulse damper)
System SoftwareChromeleon
Voltage120 to 230 VAC
Weight (Metric)6.8 kg
Flow Accuracy3%
Flow Precision0.5%
Max. Flow Rate9.99 mL/min.
Pressure Range0.35 to 17 Mpa
TypeSingle-Piston, Isocratic Metering Pump
Unit SizeEach