Dionex™ AS-HV Autosampler Probes
Actual product may vary
Thermo Scientific™
Dionex™ AS-HV Autosampler Probes
Support reliable, contamination-free fluid delivery for high-volume ion chromatography workflows using Thermo Scientific™ Dionex™ AS-HV Autosampler Probes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
064056Sample Probe Kit
064254PEEK Probe
Catalog number 064056
Price (INR)
-
Product Type:
Sample Probe Kit
Support reliable, contamination-free fluid delivery for high-volume ion chromatography workflows using Thermo Scientific™ Dionex™ AS-HV Autosampler Probes.
PEEK sample probes are designed to withstand aggressive chemicals and extreme pH levels. By utilizing an ultra-clean, completely metal-free fluid path, this probe prevents sample adsorption and eliminates trace metal leaching. It is the ideal solution for laboratories performing sensitive trace-level environmental or industrial analysis where absolute sample integrity and reproducible results are critical.
Specifications
DescriptionPTFE Lined and sleeved
For Use With (Application)Ion Chromatography
For Use With (Equipment)AS-HV Autosampler
Product TypeSample Probe Kit
Unit SizeEach