Dionex™ AS-HV Autosampler Sample Rack
Actual product may vary
Dionex™ AS-HV Autosampler Sample Rack
Dionex™ AS-HV Autosampler Sample Rack
Dionex™ AS-HV Autosampler Sample Rack
Dionex™ AS-HV Autosampler Sample Rack
Thermo Scientific™
Dionex™ AS-HV Autosampler Sample Rack
Secure racks to hold sample tubes and bottles for the Thermo Scientific™ Dionex™ AS-HV Autosampler.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
064234250 mL Nalgene Bottles
06449850 mL Vials/Tubes
06449926 mL Vials/Tubes
06450015 mL Vials/Tubes
06450110 mL Vials/Tubes
064826125 mL Nalgene Bottles
064252AS-HV Autosampler
Catalog number 064234
Price (INR)
-
For Use With (Equipment):
250 mL Nalgene Bottles
Conveniently secure dozens of sample vials, tubes or bottles for the Thermo Scientific™ Dionex™ AS-HV Autosampler. Space-saving Dionex™ AS-HV Autosampler Sample Racks are available for centrifuge tubes and Nalgene™ bottles. The AS-HV Autosampler can hold up to three racks of the same type.
Specifications
DescriptionFor 250 mL Nalgene bottles
For Use With (Application)Ion Chromatography
For Use With (Equipment)250 mL Nalgene Bottles
Product TypeAS-HV Sample Rack
Sample Capacity15 Position
Unit SizeEach