Dionex™ AS-HV Autosampler Bottles and Closures
Actual product may vary
Thermo Scientific™

Dionex™ AS-HV Autosampler Bottles and Closures

Thermo Scientific™ Dionex™ AS-HV Autosampler Bottles and Closures increase the flexibility of the AS-HV.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
064235Tissue Culture Flasks
064232Nalgene Bottles
Catalog number 064235
Price (INR)
-
Description:
Tissue Culture Flasks

Thermo Scientific™ Dionex™ AS-HV Autosampler Bottles and Closures increase the flexibility of the AS-HV.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionTissue Culture Flasks
For Use With (Application)Ion Chromatography
For Use With (Equipment)Dionex AS-HV Autosampler
QuantityPack of 50
TypeCaps and Septa
Volume (Metric)250 mL
Unit SizePack of 50