Dionex™ AS-HV Autosampler Sample Rack
Actual product may vary
Dionex™ AS-HV Autosampler Sample Rack
Dionex™ AS-HV Autosampler Sample Rack
Dionex™ AS-HV Autosampler Sample Rack
Dionex™ AS-HV Autosampler Sample Rack
Thermo Scientific™
Dionex™ AS-HV Autosampler Sample Rack
Secure racks to hold sample tubes and bottles for the Thermo Scientific™ Dionex™ AS-HV Autosampler.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
06449850 mL Vials/Tubes
06449926 mL Vials/Tubes
06450015 mL Vials/Tubes
06450110 mL Vials/Tubes
064826125 mL Nalgene Bottles
064234250 mL Nalgene Bottles
064252AS-HV Autosampler
Catalog number 064498
Price (INR)
-
For Use With (Equipment):
50 mL Vials/Tubes
Conveniently secure dozens of sample vials, tubes or bottles for the Thermo Scientific™ Dionex™ AS-HV Autosampler. Space-saving Dionex™ AS-HV Autosampler Sample Racks are available for centrifuge tubes and Nalgene™ bottles. The AS-HV Autosampler can hold up to three racks of the same type.
Specifications
For Use With (Equipment)50 mL Vials/Tubes
Product TypeAS-HV Sample Rack
Sample Capacity21 Position
Unit SizeEach