Dionex™ ASE™ 150/350 Collection Vial Inserts
Actual product may vary
Dionex™ ASE™ 150/350 Collection Vial Inserts
Dionex™ ASE™ 150/350 Collection Vial Inserts
Thermo Scientific™
Dionex™ ASE™ 150/350 Collection Vial Inserts
Use these inserts to accommodate 60mL collection vials on Thermo Scientific™ Dionex™ ASE™ 150/350 Accelerated Solvent Extractors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)
06639260 mL Vial Insert CμpDionex ASE 150
06617660 mL Vial Insert TrayDionex ASE 350
Catalog number 066392
Price (INR)
-
Description:
60 mL Vial Insert Cμp
For Use With (Equipment):
Dionex ASE 150
These inserts accommodate 60mL collection vials on Thermo Scientific™ Dionex™ ASE™ 150/350 Accelerated Solvent Extractors.
Capacity of Dionex ASE 350 Collection Tray and Vial Insert
When installed by itself, the Dionex ASE 350 Collection Tray can hold up to 19 collection bottles (250mL) and two rinse bottles. When the Dionex ASE 350 Tray Insert is installed, the collection tray can hold up to 28 collection vials (60mL) on the outside row, plus five collection bottles (250mL) and two rinse bottles on the inside row.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description60 mL Vial Insert Cμp
QuantityEach 1
For Use With (Equipment)Dionex ASE 150
TypeVial Insert Cup
Unit SizeEach