Dionex™ AS-DV Autosampler Internal Injection Port Valve Kits
Actual product may vary
Thermo Scientific™

Dionex™ AS-DV Autosampler Internal Injection Port Valve Kits

Choose high-pressure switching valves for the Thermo Scientific™ Dionex™ AS-DV Autosampler, in 6-port and 10-port configurations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
0689206-Port Valve Kit
06892110-Port Valve Kit
Catalog number 068920
Price (INR)
-
Product Type:
6-Port Valve Kit

Get the right internal injection valve for your applications, Thermo Scientific™ Dionex™ AS-DV Autosampler Port Valve Kits provide high-pressure switching valves for the AS-DV Autosampler, in a choice of 6-port and 10-port configurations.

The valves can perform the following functions:
  • Bleed Valve
  • Injection Valve
  • Matrix Elimination
  • Concentration
  • Large/Small Loop Reinjection
Specifications
For Use With (Equipment)AS-DV Autosampler
Product Type6-Port Valve Kit
Unit SizeEach