Dionex™ CRD Vacuum Mode Ship Kit
Dionex™ CRD Vacuum Mode Ship Kit
Actual product may vary
Thermo Scientific™
Dionex™ CRD Vacuum Mode Ship Kit
Use the Thermo Scientific™ Dionex™ CRD Vacuum Mode Ship Kit to enable operation of Dionex CRD 200 and CRD 300 Carbonate Removal Devices in vacuum mode to support specific applications such as analysis of anions in an ammoniated water sample.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
069414Tubing
064911Vacuum Mode Ship Kit
Catalog number 069414
Price (INR)
-
Request A Quote
Type:
Tubing
Use the Thermo Scientific™ Dionex™ CRD Vacuum Mode Ship Kit to enable operation of Dionex CRD 200 and CRD 300 Carbonate Removal Devices in vacuum mode to support specific applications such as analysis of anions in an ammoniated water sample.
Carbonate Removal Device Accessories
  • Santoprene Tubing for Peristaltic Pump
  • Ship Kit for Peristaltic Pump/Dionex CRD 300
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Equipment)Perstaltic Pump for The Dionex CRD 300
IncludesSantoprene tubing for Peristaltic Pump (30 cm)
QuantityEach 1
TypeTubing
Unit SizeEach 1