Dionex™ AS-AP Autosampler Valves
Actual product may vary
Thermo Scientific™

Dionex™ AS-AP Autosampler Valves

Thermo Scientific™ Dionex™ AS-AP Autosampler Valves are high-pressure switching valves, available in 6-port and 10-port configurations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeIncludes
070208Secondary Valve2-way, 10-port valve
070209Secondary Valve2 way 6-port valve
074122Valve Assembly2-way, 10-port Valve, Mounting Hardware
074123Valve Assembly2-way, 6-port Valve, Mounting Hardware
Catalog number 070208
Price (INR)
-
Type:
Secondary Valve
Includes:
2-way, 10-port valve

Thermo Scientific™ Dionex™ AS-AP Autosampler Valves are high-pressure switching valves, available in 6-port and 10-port configurations.

The valves can perform the following functions:

  • Bleed Valve
  • Injection Valve
  • Matrix Elimination
  • Concentration
  • Large/Small Loop Reinjection
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionSecondary 2-way, 10-port valve
For Use With (Equipment)AS-AP Autosampler
Includes2-way, 10-port valve
QuantityEach 1
TypeSecondary Valve
Unit SizeEach 1