Dionex™ ICS-6000 Carbohydrate Removal Accessory Kit for ED Electrochemical Detection
Actual product may vary
Thermo Scientific™
Dionex™ ICS-6000 Carbohydrate Removal Accessory Kit for ED Electrochemical Detection
Save time and improve amino acid analyses by removing carbohydrates in-line, using Thermo Scientific™ Dionex™ ICS-6000 Carbohydrate Removal Accessory Kit for ED Electrochemical Detection.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
070510Carbohydrate Removal SP ED Detector
063522Carbohydrate Removal ED Detector
Catalog number 070510
Price (INR)
-
Description:
Carbohydrate Removal SP ED Detector
Eliminate labor-intensive sample cleanup and ensure reliable amino acid quantification by removing carbohydrates from amino acid samples in-line using the Thermo Scientific™ Dionex™ ICS-6000 Carbohydrate Removal Accessory Kits for ED Electrochemical Detection. The syringe pump and six-port valve in this kit work in conjunction with Thermo Scientific™ Dionex™ CRC Carbohydrate Removal Cartridges to enable automatic pretreatment of your amino acid samples on a Thermo Scientific™ Dionex™ AAA-Direct™ ion chromatography system.
The kit's syringe pump and six-port valve is designed for the Thermo Scientific™ Dionex™ ICS-5000+/ICS-6000 DP Dual Pump, or the Dionex ICS-5000+/ICS-6000 SP Single Pump system and a Dionex Carbohydrate Removal Cartridge. Installation requires modification of the Dionex AAA-Direct system to connect the syringe pump to the autosampler, and redirect the pump and Dionex™ AminoPac PA™ 10 to the six-port switching valve.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionCarbohydrate Removal SP ED Detector
For Use With (Equipment)ICS-5000+/ICS-6000 SP ED Detectors
IncludesSyringe pump and six-port valve
OperationModifies AAA-Direct system to accommodate a Carbohydrate Removal Cartridge.
QuantityEach 1
TypeCarbohydrate Removal Accessory Kit
Unit SizeEach 1