Dionex™ EO Eluent Organizer Tray
Thermo Scientific™
Dionex™ EO Eluent Organizer Tray
Organize your eluent, tubing, and air lines in one convenient, leak-proof caddy.
Have Questions?
Catalog NumberIncludes
072059EO Accessory Tray Only
Catalog number 072059
Price (INR)
-
Includes:
EO Accessory Tray Only
Organize your eluent, tubing, and air lines in one convenient, leak- and corrosion-proof caddy. The Thermo Scientific™ Dionex™ EO Eluent Organizer Tray holds two to four, glass or plastic Dionex™ EO Eluent Organizer Bottles and optional Dionex™ EO Eluent Organizer Regulator Kit (bottles and gas regulator sold separately).
Sturdy Storage
Leak-free translucent liner
Corrosion-proof polypropylene and epoxy construction
Large capacity:
four, 1–2L plastic eluent bottles;
two, 1L glass eluent bottles; or
one, 2L glass eluent bottle
Space-saving design
Up to two Dionex™ EO Eluent Organizers stack on the top of one Dionex™ DC Detector/Chromatography Compartment or Dionex™ TC Thermal Compartment (TC) module. Use the Dionex™ EO Eluent Organizer Regulator Accessory and Stand to securely mount the eluent organizer and gas regulator onto the detector.
Specifications
DescriptionTray only
For Use With (Equipment)EO Eluent Organizer Bottles, EO Eluent Organizer Regulator Kit
IncludesEO Accessory Tray Only
OperationContains spills and allows view of liquid levels.
TypeEluent Organizer Tray
Unit SizeEach