Dionex™ AutoTrace™ 280 Solid-Phase Extraction Instrument
Actual product may vary
Dionex™ AutoTrace™ 280 Solid-Phase Extraction Instrument
Dionex™ AutoTrace™ 280 Solid-Phase Extraction Instrument
Dionex™ AutoTrace™ 280 Solid-Phase Extraction Instrument
Thermo Scientific™

Dionex™ AutoTrace™ 280 Solid-Phase Extraction Instrument

Green features

Reduce labor and solvent costs by up to 90% with automated solid-phase extraction system.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
072606Cartridge with 6 mL Glass Plungers
071385Cartridges with 6 mL Plungers
07138647 mm Disks
072604Cartridges with 1 mL Plungers
072605Cartridge with 3 mL Plungers
Catalog number 072606
Price (INR)
-
For Use With (Equipment):
Cartridge with 6 mL Glass Plungers

Extract analytes of interest from large volume aqueous samples easily and efficiently with the Thermo Scientific™ Dionex™ AutoTrace™ 280 Solid-Phase Extraction (SPE) Instrument. The AutoTrace 280 automates all four steps of SPE (conditioning, loading, rinsing, and eluting), reducing solvent consumption and improving recovery and reproducibility. Boost productivity and reduce cost of analysis by simultaneously processing up to six samples in 2-3 hours with only 15 minutes of operator involvement. The AutoTrace 280 meets requirements in US EPA methods including the 500 and 600 series.

Improved reliability and recovery
  • Automated SPE minimizes operator intervention
  • Positive pressure loading and elution of samples and solvents provide constant flow for improved analytical precision

Flexibility in operation
  • Loads sample volumes from 20 mL to 4 L
  • Compatible with 1, 3, and 6 mL cartridges and with 47 mm SPE disks
  • Automatically conditions, rinses, and elutes SPE cartridges with a choice of five solvents
  • Offers a choice of six different collection vial racks

Improved laboratory safety
  • Minimal sample handling reduces operator exposure to hazardous chemicals
  • No hood required

Enhanced laboratory productivity and reduced costs
  • Unattended operation
  • Batch processing of up to 6 samples
  • Reduced solvent usage
  • Segregates aqueous and solvent waste using separate waste lines for efficient, cost-effective waste handling

Supports a range of matrices and applications
  • Ability to process samples containing suspended solids
  • Meets requirements for solid-phase extraction for U.S. EPA methods including 500 and 600 series

Recommended use
  • Analyze prepared samples with Thermo Scientific LC or GC systems for a fully integrated workflow
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionFor cartridges with 6 mL glass plungers
Frequency47 to 63 Hz
PressureOutput: 0 to 30 psi (0 to 1.4 bars), Input: 100 psi (6.9 bars) maximum
Dimensions (D x W x H)Instrument: 57 x 63.5 x 69 cm
For Use With (Equipment)Cartridge with 6 mL Glass Plungers
TypeAutoTrace 280 SPE System
Voltage100/240 V ±10%
Unit SizeEach