Dionex™ AS-AP Autosampler Sample Syringes
Actual product may vary
Thermo Scientific™
Dionex™ AS-AP Autosampler Sample Syringes
Employ high-quality syringes for the Thermo Scientific™ Dionex™ AS-AP Autosampler.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
0743085 mL
074305100 μL
074306250 μL
0743071000 μL
Catalog number 074308
Price (INR)
-
Volume (Metric):
5 mL
To suit your sample and application needs, sample syringes for the Thermo Scientific™ Dionex™ AS-AP Autosampler are available in four convenient sizes: 100μL, 250μL, 1000μL and 5mL.
Specifications
For Use With (Equipment)AS-AP Autosampler
Product TypeSample Syringe
Volume (Metric)5 mL
Unit SizeEach