Dionex™ SolEx™ Carbon-Based SPE Cartridges for EPA Method 521, 522, and 535
Actual product may vary
Dionex™ SolEx™ Carbon-Based SPE Cartridges for EPA Method 521, 522, and 535
Dionex™ SolEx™ Carbon-Based SPE Cartridges for EPA Method 521, 522, and 535
Dionex™ SolEx™ Carbon-Based SPE Cartridges for EPA Method 521, 522, and 535
Thermo Scientific™
Dionex™ SolEx™ Carbon-Based SPE Cartridges for EPA Method 521, 522, and 535
Use these cartridges for fast and easy SPE of nitrosamines (EPA Method 521), 1,4-dioxane (EPA Method 522) and herbicides (EPA Method 523) in drinking water.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Application)
075898EPA Method 535
074590US EPA Method 521 and 522
Catalog number 075898
Price (INR)
-
For Use With (Application):
EPA Method 535
For fast and easy solid-phase extraction (SPE) of nitrosamines (U.S. EPA Method 521) and 1,4-dioxane (U.S. EPA Method 522) in drinking water, use the Thermo Scientific™ Dionex™ SolEx™ Carbon-Based SPE Cartridges. This cartridge is available in 6mL format with 2g of packing. For SPE of herbicides (U.S. EPA Method 535) in drinking water, use the Thermo Scientific™ Dionex™ SolEx™ Graphitized Carbon SPE Cartridge. This cartridge is available in 6mL formats with 0.5g of packing. These cartridges are designed for use with the Thermo Scientific™ Dionex™ AutoTrace™ 280 SPE system or other SPE instrumentation such as vacuum manifolds.
Carbon Activated Carbon (charcoal)
  • Particle Size: 972 m2/g
Graphitized Carbon
  • Non-Porous Graphitized Carbon Particles
  • Particle Size: 90 m2/g
About Dionex SolEx SPE Cartridges
SolEx SPE Cartridges extract organic pollutants from a liquid sample off-line using a solid-phase adsorbent. The cartridges employ a variety of chemistries depending on the application. The silica- or carbon-based absorbents are bound or encapsulated in a membrane filter (called disk format) or packed in a cartridge (syringe bodies). Water (ground, waste, and drinking) is the most common liquid sample extracted, but these SPE cartridges are also used to extract contaminants from milk, fruit juices, urine, and other matrices. These cartridges are designed for use with the AutoTrace 280 SPE system or other SPE instrumentation such as vacuum manifolds.
  • Sorbant material made of irregular bonded silica particles
  • Excellent capacity from packed with 40 to 63 micron particles
  • Long 5-year shelf life
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description6 mL; 0.5 g of packing
For Use With (Application)EPA Method 535
QuantityPack of 30
For Use With (Equipment)AutoTrace 280 System
SorbentCarbon-535
TypeGraphitized Carbon SPE Cartridge
Unit SizePack of 30