Dionex™ IonPac™ Mixer
Thermo Scientific™
Dionex™ IonPac™ Mixer
Pair this mixer with the Thermo Scientific™ Dionex™ SCS 1 Silica Cation Separator column for non-suppressed conductivity detection or single-column ion chromatography (SCIC).
Have Questions?
Catalog NumberWeight (Metric)
0799710.070 kg
Catalog number 079971
Price (INR)
-
Weight (Metric):
0.070 kg
Reduce background noise for non-suppressed conductivity detection, or SCIC, analysis with the Thermo Scientific™ Dionex™ IonPac™ Mixer. The Dionex IonPac Mixer is required for use with the Dionex SCS 1 column to average any eluent concentration changes due to temperature fluctuation or pump pulsations. The mixer should be installed before the eluent inlet of the injection valve.
  • Required for use with Dionex SCS 1 column.
Specifications
Capacity500 μL
For Use With (Equipment)Dionex IonPac SCS 1 Silica Cation Separator Column
Volume (Metric)1.087 L
Weight (Metric)0.070 kg
Unit SizeEach