Orion™ BOD Adapters for Polarographic Dissolved Oxygen Probes
Thermo Scientific™

Orion™ BOD Adapters for Polarographic Dissolved Oxygen Probes

Help ensure proper and optimal probe performance with the Thermo Scientific™ Orion™ BOD Adapters for Polarographic Dissolved Oxygen Probes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
080360Orion 830A Series DO Probes
080160Orion 810 Series DO Probes
Catalog number 080360
Price (INR)
-
For Use With (Equipment):
Orion 830A Series DO Probes

Help ensure proper and optimal probe performance with Thermo Scientific™ Orion™ BOD Adapters for Polarographic Dissolved Oxygen Probes. Adapters include stir funnel and electrode adapter sleeve for BOD bottle.

Warranty: Out of box.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionFor use with 083005MD, 083010MD, 083025MD, 083060MD, 083005A, 083010A, 083005
IncludesStir funnel and electrode adaptor sleeve
For Use With (Equipment)Orion 830A Series DO Probes
TypeBOD Adapter
Unit SizeEach